Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602835 | ||||||||||||||||||||
Literature Links: |
GAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GAS2 - growth arrest specific 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143830.1 | Intron | NP_001137302.1 | ||||
NM_005256.3 | Intron | NP_005247.1 | ||||
NM_177553.2 | Intron | NP_808221.1 | ||||
XM_011519971.2 | Intron | XP_011518273.1 | ||||
XM_011519972.2 | Intron | XP_011518274.1 | ||||
XM_011519975.1 | Intron | XP_011518277.1 | ||||
XM_011519978.2 | Intron | XP_011518280.1 | ||||
XM_017017528.1 | Intron | XP_016873017.1 | ||||
XM_017017529.1 | Intron | XP_016873018.1 | ||||
XM_017017530.1 | Intron | XP_016873019.1 | ||||
XM_017017531.1 | Intron | XP_016873020.1 | ||||
XM_017017532.1 | Intron | XP_016873021.1 |
SVIP - small VCP interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320340.1 | Intron | NP_001307269.1 | ||||
NM_001320341.1 | Intron | NP_001307270.1 | ||||
NM_001320342.1 | Intron | NP_001307271.1 | ||||
NM_148893.2 | Intron | NP_683691.1 |