Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||
Phenotype: |
MIM: 605722 | ||||||||||||||||||||||||||
Literature Links: |
KCNJ16 PubMed Links | ||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
KCNJ16 - potassium voltage-gated channel subfamily J member 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270422.1 | Intron | NP_001257351.1 | ||||
NM_001291622.1 | Intron | NP_001278551.1 | ||||
NM_001291623.1 | Intron | NP_001278552.1 | ||||
NM_001291624.1 | Intron | NP_001278553.1 | ||||
NM_001291625.1 | Intron | NP_001278554.1 | ||||
NM_018658.2 | Intron | NP_061128.2 | ||||
NM_170741.2 | Intron | NP_733937.2 | ||||
NM_170742.2 | Intron | NP_733938.2 | ||||
XM_005257337.4 | Intron | XP_005257394.1 | ||||
XM_006721885.3 | Intron | XP_006721948.1 | ||||
XM_006721886.3 | Intron | XP_006721949.1 | ||||
XM_006721887.3 | Intron | XP_006721950.1 | ||||
XM_011524781.2 | Intron | XP_011523083.1 | ||||
XM_017024609.1 | Intron | XP_016880098.1 | ||||
XM_017024610.1 | Intron | XP_016880099.1 | ||||
XM_017024611.1 | Intron | XP_016880100.1 | ||||
XM_017024612.1 | Intron | XP_016880101.1 | ||||
XM_017024613.1 | Intron | XP_016880102.1 |
LINC01028 - long intergenic non-protein coding RNA 1028 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |