Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608704 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NMRK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NMRK1 - nicotinamide riboside kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127603.1 | 1937 | UTR 3 | NP_001121075.1 | |||
NM_017881.2 | 1937 | UTR 3 | NP_060351.1 | |||
XM_006717162.1 | 1937 | UTR 3 | XP_006717225.1 | |||
XM_006717163.2 | 1937 | UTR 3 | XP_006717226.1 | |||
XM_011518817.1 | 1937 | UTR 3 | XP_011517119.1 | |||
XM_011518818.1 | 1937 | UTR 3 | XP_011517120.1 | |||
XM_011518819.2 | 1937 | UTR 3 | XP_011517121.1 | |||
XM_017014868.1 | 1937 | UTR 3 | XP_016870357.1 | |||
XM_017014869.1 | 1937 | UTR 3 | XP_016870358.1 | |||
XM_017014870.1 | 1937 | UTR 3 | XP_016870359.1 | |||
XM_017014871.1 | 1937 | UTR 3 | XP_016870360.1 | |||
XM_017014872.1 | 1937 | UTR 3 | XP_016870361.1 | |||
XM_017014873.1 | 1937 | UTR 3 | XP_016870362.1 | |||
XM_017014874.1 | 1937 | UTR 3 | XP_016870363.1 |
Set Membership: |
HapMap |