Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RALGAPB PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RALGAPB - Ral GTPase activating protein non-catalytic beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282917.1 | Intron | NP_001269846.1 | ||||
NM_001282918.1 | Intron | NP_001269847.1 | ||||
NM_020336.3 | Intron | NP_065069.1 | ||||
XM_005260462.2 | Intron | XP_005260519.1 | ||||
XM_005260463.2 | Intron | XP_005260520.1 | ||||
XM_005260464.2 | Intron | XP_005260521.1 | ||||
XM_005260465.2 | Intron | XP_005260522.1 | ||||
XM_017027966.1 | Intron | XP_016883455.1 | ||||
XM_017027967.1 | Intron | XP_016883456.1 | ||||
XM_017027968.1 | Intron | XP_016883457.1 | ||||
XM_017027969.1 | Intron | XP_016883458.1 | ||||
XM_017027970.1 | Intron | XP_016883459.1 |