Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACCACTCAATAGGCAGCTTATTA[A/T]TTTCCTATTTACATACAAGACAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 142409 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HGF PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
HGF - hepatocyte growth factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000601.5 | 5830 | UTR 3 | NP_000592.3 | |||
NM_001010931.2 | 5830 | Intron | NP_001010931.1 | |||
NM_001010932.2 | 5830 | UTR 3 | NP_001010932.1 | |||
NM_001010933.2 | 5830 | Intron | NP_001010933.1 | |||
NM_001010934.2 | 5830 | Intron | NP_001010934.1 | |||
XM_006715956.2 | 5830 | UTR 3 | XP_006716019.1 | |||
XM_011516115.2 | 5830 | Intron | XP_011514417.1 | |||
XM_017012097.1 | 5830 | Intron | XP_016867586.1 | |||
XM_017012098.1 | 5830 | Intron | XP_016867587.1 |
LOC100128317 - uncharacterized LOC100128317 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |