Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTACTTCCTGCTGTACTTGAATAT[G/T]GGCTTCCTGCCCTAGGAAGGAAGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SYTL3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU) - Not Available | ||||||
EAS
|
African American
|
YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SYTL3 - synaptotagmin like 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001009991.3 | Intron | NP_001009991.2 | ||||
NM_001242384.1 | Intron | NP_001229313.1 | ||||
NM_001242394.1 | Intron | NP_001229323.1 | ||||
NM_001242395.1 | Intron | NP_001229324.1 | ||||
NM_001318745.1 | Intron | NP_001305674.1 | ||||
XM_005267215.4 | Intron | XP_005267272.1 | ||||
XM_005267218.4 | Intron | XP_005267275.1 | ||||
XM_005267222.4 | Intron | XP_005267279.1 | ||||
XM_006715605.3 | Intron | XP_006715668.1 | ||||
XM_006715606.3 | Intron | XP_006715669.1 | ||||
XM_006715611.3 | Intron | XP_006715674.1 | ||||
XM_011536254.2 | Intron | XP_011534556.1 | ||||
XM_011536255.2 | Intron | XP_011534557.1 | ||||
XM_017011495.1 | Intron | XP_016866984.1 | ||||
XM_017011496.1 | Intron | XP_016866985.1 | ||||
XM_017011497.1 | Intron | XP_016866986.1 | ||||
XM_017011498.1 | Intron | XP_016866987.1 | ||||
XM_017011499.1 | Intron | XP_016866988.1 |
Set Membership: |
Validated |