Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAAACCCTCGGGGAAGGCGGCAC[G/T]GACGCCCAGGCCTCTGTCCATGGGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 151750 | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPE PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LIPE - lipase E, hormone sensitive type | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005357.3 | 1954 | Missense Mutation | AGT,CGT | S,R 938 | NP_005348.2 | |
XM_005258937.3 | 1954 | Missense Mutation | AGT,CGT | S,R 862 | XP_005258994.1 | |
XM_005258938.4 | 1954 | Missense Mutation | AGT,CGT | S,R 683 | XP_005258995.1 | |
XM_005258939.3 | 1954 | Missense Mutation | AGT,CGT | S,R 700 | XP_005258996.2 | |
XM_005258940.3 | 1954 | Missense Mutation | AGT,CGT | S,R 637 | XP_005258997.1 | |
XM_006723218.2 | 1954 | Missense Mutation | AGT,CGT | S,R 637 | XP_006723281.1 | |
XM_017026810.1 | 1954 | Missense Mutation | AGT,CGT | S,R 637 | XP_016882299.1 |
LIPE-AS1 - LIPE antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC101930071 - uncharacterized LOC101930071 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |