Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613331 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MARCH1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
MARCH1 - membrane associated ring-CH-type finger 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001166373.1 | 3836 | UTR 3 | NP_001159845.1 | |||
NM_017923.3 | 3836 | UTR 3 | NP_060393.1 | |||
XM_011532055.2 | 3836 | UTR 3 | XP_011530357.1 | |||
XM_017008334.1 | 3836 | UTR 3 | XP_016863823.1 | |||
XM_017008335.1 | 3836 | UTR 3 | XP_016863824.1 | |||
XM_017008336.1 | 3836 | UTR 3 | XP_016863825.1 |
TMA16 - translation machinery associated 16 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |