Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATAGAGGTGAAGCCTGAATGAACA[C/T]GTGTCCTTTCCTTTTTTAGGTGACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609071 MIM: 604452 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXW2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
FBXW2 - F-box and WD repeat domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC100288842 - UDP-GlcNAc:betaGal beta-1,3-N-acetylglucosaminyltransferase 5 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSMD5 - proteasome 26S subunit, non-ATPase 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270427.1 | Intron | NP_001257356.1 | ||||
NM_005047.3 | Intron | NP_005038.1 | ||||
XM_011518866.2 | Intron | XP_011517168.1 | ||||
XM_017014927.1 | Intron | XP_016870416.1 | ||||
XM_017014928.1 | Intron | XP_016870417.1 |
Set Membership: |
HapMap Validated |