Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614362 MIM: 601149 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ACSBG1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ACSBG1 - acyl-CoA synthetase bubblegum family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199377.1 | 2669 | UTR 3 | NP_001186306.1 | |||
NM_015162.4 | 2669 | UTR 3 | NP_055977.3 | |||
XM_011521390.2 | 2669 | UTR 3 | XP_011519692.1 | |||
XM_011521391.2 | 2669 | Intron | XP_011519693.2 | |||
XM_017022025.1 | 2669 | Intron | XP_016877514.1 | |||
XM_017022026.1 | 2669 | UTR 3 | XP_016877515.1 |
IDH3A - isocitrate dehydrogenase 3 (NAD(+)) alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005530.2 | 2669 | Intron | NP_005521.1 | |||
XM_005254336.2 | 2669 | Intron | XP_005254393.1 | |||
XM_017022118.1 | 2669 | Intron | XP_016877607.1 | |||
XM_017022119.1 | 2669 | Intron | XP_016877608.1 | |||
XM_017022120.1 | 2669 | Intron | XP_016877609.1 |
Set Membership: |
HapMap |