Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611482 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKRD37 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANKRD37 - ankyrin repeat domain 37 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_181726.3 | 2006 | Intron | NP_859077.1 | |||
XM_017008176.1 | 2006 | UTR 3 | XP_016863665.1 |
C4orf47 - chromosome 4 open reading frame 47 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LRP2BP - LRP2 binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UFSP2 - UFM1 specific peptidase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018359.3 | 2006 | UTR 3 | NP_060829.2 |