Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 157680 MIM: 609372 | ||||||||||||||||||||
Literature Links: |
CDC25C PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDC25C - cell division cycle 25C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287582.1 | Intron | NP_001274511.1 | ||||
NM_001287583.1 | Intron | NP_001274512.1 | ||||
NM_001318098.1 | Intron | NP_001305027.1 | ||||
NM_001790.4 | Intron | NP_001781.2 | ||||
NM_022809.3 | Intron | NP_073720.1 | ||||
XM_005272145.3 | Intron | XP_005272202.1 | ||||
XM_006714739.3 | Intron | XP_006714802.1 | ||||
XM_011543759.2 | Intron | XP_011542061.1 | ||||
XM_011543760.2 | Intron | XP_011542062.1 | ||||
XM_011543761.2 | Intron | XP_011542063.1 | ||||
XM_011543762.1 | Intron | XP_011542064.1 | ||||
XM_011543763.1 | Intron | XP_011542065.1 | ||||
XM_011543764.1 | Intron | XP_011542066.1 | ||||
XM_017010101.1 | Intron | XP_016865590.1 |
FAM53C - family with sequence similarity 53 member C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135647.1 | Intron | NP_001129119.1 | ||||
NM_016605.2 | Intron | NP_057689.1 |