Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTTGACTACCAGACTATCATGAT[A/G]TTTGTTGGAACTGTAATTCCTGCTC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 616305 MIM: 604993 | ||||||||||||||||||||
Literature Links: |
FRMD4A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FRMD4A - FERM domain containing 4A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318336.1 | 6099 | UTR 3 | NP_001305265.1 | |||
NM_001318337.1 | 6099 | UTR 3 | NP_001305266.1 | |||
NM_001318338.1 | 6099 | UTR 3 | NP_001305267.1 | |||
NM_018027.4 | 6099 | UTR 3 | NP_060497.3 | |||
XM_005252489.2 | 6099 | Intron | XP_005252546.1 | |||
XM_005252490.2 | 6099 | Intron | XP_005252547.1 | |||
XM_005252495.4 | 6099 | Intron | XP_005252552.1 | |||
XM_006717457.2 | 6099 | Intron | XP_006717520.1 | |||
XM_006717458.3 | 6099 | Intron | XP_006717521.1 | |||
XM_006717459.2 | 6099 | Intron | XP_006717522.1 | |||
XM_006717460.2 | 6099 | Intron | XP_006717523.1 | |||
XM_006717464.1 | 6099 | Intron | XP_006717527.1 | |||
XM_011519539.1 | 6099 | Intron | XP_011517841.1 | |||
XM_011519540.1 | 6099 | Intron | XP_011517842.1 | |||
XM_011519541.1 | 6099 | Intron | XP_011517843.1 | |||
XM_011519543.2 | 6099 | Intron | XP_011517845.1 | |||
XM_017016393.1 | 6099 | Intron | XP_016871882.1 | |||
XM_017016394.1 | 6099 | Intron | XP_016871883.1 | |||
XM_017016395.1 | 6099 | Intron | XP_016871884.1 | |||
XM_017016396.1 | 6099 | Intron | XP_016871885.1 |
LOC101928524 - uncharacterized LOC101928524 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRPF18 - pre-mRNA processing factor 18 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003675.3 | 6099 | Intron | NP_003666.1 | |||
XM_011519754.2 | 6099 | Intron | XP_011518056.1 | |||
XM_017016860.1 | 6099 | Intron | XP_016872349.1 |