Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608009 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AK5 - adenylate kinase 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
ZZZ3 - zinc finger ZZ-type containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308237.1 | 1566 | Silent Mutation | CAA,CAG | Q,Q 374 | NP_001295166.1 | |
NM_015534.5 | 1566 | Silent Mutation | CAA,CAG | Q,Q 868 | NP_056349.1 | |
XM_005270725.3 | 1566 | Silent Mutation | CAA,CAG | Q,Q 868 | XP_005270782.1 | |
XM_005270726.3 | 1566 | Silent Mutation | CAA,CAG | Q,Q 868 | XP_005270783.1 | |
XM_005270727.3 | 1566 | Silent Mutation | CAA,CAG | Q,Q 867 | XP_005270784.1 | |
XM_005270729.4 | 1566 | Silent Mutation | CAA,CAG | Q,Q 374 | XP_005270786.1 | |
XM_017000970.1 | 1566 | Silent Mutation | CAA,CAG | Q,Q 867 | XP_016856459.1 | |
XM_017000971.1 | 1566 | Intron | XP_016856460.1 | |||
XM_017000972.1 | 1566 | Silent Mutation | CAA,CAG | Q,Q 373 | XP_016856461.1 |