Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRRIQ3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
LRRIQ3 - leucine rich repeats and IQ motif containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001105659.1 | Intron | NP_001099129.1 | ||||
NM_001322315.1 | Intron | NP_001309244.1 | ||||
XM_011540658.1 | Intron | XP_011538960.1 | ||||
XM_011540659.1 | Intron | XP_011538961.1 | ||||
XM_011540660.1 | Intron | XP_011538962.1 | ||||
XM_011540662.2 | Intron | XP_011538964.1 | ||||
XM_011540663.2 | Intron | XP_011538965.1 | ||||
XM_017000286.1 | Intron | XP_016855775.1 | ||||
XM_017000287.1 | Intron | XP_016855776.1 | ||||
XM_017000288.1 | Intron | XP_016855777.1 |
Set Membership: |
HapMap Validated |