Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCTTTCTACCTCTACCACAGATG[C/G]ATTTATGAAATGTTGATCAATGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616889 MIM: 179508 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP68 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP68 - centrosomal protein 68 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319100.1 | 1669 | Intron | NP_001306029.1 | |||
NM_001319101.1 | 1669 | Intron | NP_001306030.1 | |||
NM_015147.2 | 1669 | Intron | NP_055962.2 | |||
XM_005264219.3 | 1669 | Intron | XP_005264276.1 | |||
XM_017003633.1 | 1669 | Intron | XP_016859122.1 |
RAB1A - RAB1A, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004161.4 | 1669 | UTR 3 | NP_004152.1 | |||
NM_015543.1 | 1669 | UTR 3 | NP_056358.1 | |||
XM_005264468.1 | 1669 | UTR 3 | XP_005264525.1 |