Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTCTTCCGGGTATTCCTCTTCCTC[A/G]CAGGAAGCTAAGGGCATCCACGACC
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ISY1-RAB43 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ISY1-RAB43 - ISY1-RAB43 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204890.1 | Intron | NP_001191819.1 |
RAB43 - RAB43, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204883.1 | Intron | NP_001191812.1 | ||||
NM_001204884.1 | Intron | NP_001191813.1 | ||||
NM_001204885.1 | Intron | NP_001191814.1 | ||||
NM_001204886.1 | Intron | NP_001191815.1 | ||||
NM_001204887.1 | Intron | NP_001191816.1 | ||||
NM_001204888.1 | Intron | NP_001191817.1 | ||||
NM_198490.2 | Intron | NP_940892.1 |
Set Membership: |
HapMap |