Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAATGGTGAAATTGATAATTCAAA[A/G]AAATTAGCGCTGGAAGAAACAACGG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
25 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604718 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC101929099 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
LOC101929099 - uncharacterized LOC101929099 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TTF2 - transcription termination factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003594.3 | Intron | NP_003585.3 | ||||
XM_005271277.3 | Intron | XP_005271334.1 | ||||
XM_011542303.2 | Intron | XP_011540605.1 | ||||
XM_017002549.1 | Intron | XP_016858038.1 | ||||
XM_017002550.1 | Intron | XP_016858039.1 | ||||
XM_017002551.1 | Intron | XP_016858040.1 | ||||
XM_017002552.1 | Intron | XP_016858041.1 | ||||
XM_017002553.1 | Intron | XP_016858042.1 | ||||
XM_017002554.1 | Intron | XP_016858043.1 | ||||
XM_017002555.1 | Intron | XP_016858044.1 |