Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 180980 MIM: 614720 | ||||||||||||||||||||
Literature Links: |
AMD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
AMD1 - adenosylmethionine decarboxylase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001287214.1 | Intron | NP_001274143.1 | ||||
NM_001287215.1 | Intron | NP_001274144.1 | ||||
NM_001287216.1 | Intron | NP_001274145.1 | ||||
NM_001634.5 | Intron | NP_001625.2 |
CDK19 - cyclin dependent kinase 19 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |