Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107710 MIM: 107741 MIM: 608061 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
APOC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
APOC1 - apolipoprotein C1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
APOE - apolipoprotein E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000041.3 | Intron | NP_000032.1 | ||||
NM_001302688.1 | Intron | NP_001289617.1 | ||||
NM_001302689.1 | Intron | NP_001289618.1 | ||||
NM_001302690.1 | Intron | NP_001289619.1 | ||||
NM_001302691.1 | Intron | NP_001289620.1 |
TOMM40 - translocase of outer mitochondrial membrane 40 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |