Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601958 MIM: 612172 | ||||||||||||||||||||
Literature Links: |
CACNB3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CACNB3 - calcium voltage-gated channel auxiliary subunit beta 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000725.3 | 289 | Missense Mutation | GAG,GTG | E,V 60 | NP_000716.2 | |
NM_001206915.1 | 289 | Intron | NP_001193844.1 | |||
NM_001206916.1 | 289 | Missense Mutation | GAG,GTG | E,V 59 | NP_001193845.1 | |
NM_001206917.1 | 289 | Missense Mutation | GAG,GTG | E,V 47 | NP_001193846.1 | |
XM_011538727.2 | 289 | UTR 5 | XP_011537029.1 | |||
XM_011538728.2 | 289 | UTR 5 | XP_011537030.1 | |||
XM_017019957.1 | 289 | UTR 5 | XP_016875446.1 | |||
XM_017019958.1 | 289 | UTR 5 | XP_016875447.1 | |||
XM_017019959.1 | 289 | UTR 5 | XP_016875448.1 |
DDX23 - DEAD-box helicase 23 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |