Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609484 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
LYPD3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
LYPD3 - LY6/PLAUR domain containing 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PHLDB3 - pleckstrin homology like domain family B member 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_198850.3 | 2327 | Silent Mutation | ACA,ACC | T,T 632 | NP_942147.3 | |
XM_005259169.3 | 2327 | Silent Mutation | ACA,ACC | T,T 627 | XP_005259226.1 | |
XM_005259170.3 | 2327 | Silent Mutation | ACA,ACC | T,T 593 | XP_005259227.1 | |
XM_005259171.3 | 2327 | Intron | XP_005259228.1 | |||
XM_011527236.2 | 2327 | Silent Mutation | ACA,ACC | T,T 627 | XP_011525538.1 |