Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
74 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194631 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZNF124 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ZNF124 - zinc finger protein 124 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243740.2 | Intron | NP_001230669.1 | ||||
NM_001297567.1 | Intron | NP_001284496.1 | ||||
NM_001297568.1 | Intron | NP_001284497.1 | ||||
NM_001297569.1 | Intron | NP_001284498.1 | ||||
NM_003431.3 | Intron | NP_003422.2 | ||||
XM_005273257.4 | Intron | XP_005273314.1 | ||||
XM_006711813.3 | Intron | XP_006711876.1 | ||||
XM_011544272.2 | Intron | XP_011542574.1 |
Set Membership: |
HapMap Validated |