Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114190 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CALCRL PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CALCRL - calcitonin receptor like receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001271751.1 | 2547 | UTR 3 | NP_001258680.1 | |||
NM_005795.5 | 2547 | UTR 3 | NP_005786.1 | |||
XM_005246231.3 | 2547 | UTR 3 | XP_005246288.2 | |||
XM_005246232.3 | 2547 | UTR 3 | XP_005246289.2 | |||
XM_005246234.3 | 2547 | UTR 3 | XP_005246291.2 | |||
XM_017003151.1 | 2547 | UTR 3 | XP_016858640.1 |
Set Membership: |
HapMap |