Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602633 | ||||||||||||||||||||
Literature Links: |
C2orf49 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C2orf49 - chromosome 2 open reading frame 49 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286537.1 | 1412 | Intron | NP_001273466.1 | |||
NM_024093.2 | 1412 | Intron | NP_076998.1 | |||
XM_017004892.1 | 1412 | Intron | XP_016860381.1 |
FHL2 - four and a half LIM domains 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039492.2 | 1412 | UTR 3 | NP_001034581.1 | |||
NM_001318894.1 | 1412 | UTR 3 | NP_001305823.1 | |||
NM_001318895.1 | 1412 | UTR 3 | NP_001305824.1 | |||
NM_001318896.1 | 1412 | UTR 3 | NP_001305825.1 | |||
NM_001318897.1 | 1412 | UTR 3 | NP_001305826.1 | |||
NM_001318898.1 | 1412 | UTR 3 | NP_001305827.1 | |||
NM_001318899.1 | 1412 | UTR 3 | NP_001305828.1 | |||
NM_001450.3 | 1412 | UTR 3 | NP_001441.4 | |||
NM_201555.1 | 1412 | UTR 3 | NP_963849.1 | |||
NM_201557.3 | 1412 | UTR 3 | NP_963851.2 | |||
XM_011510798.2 | 1412 | UTR 3 | XP_011509100.1 |