Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 107940 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARRB1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARRB1 - arrestin beta 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004041.4 | 4156 | UTR 3 | NP_004032.2 | |||
NM_020251.3 | 4156 | UTR 3 | NP_064647.1 | |||
XM_011545034.2 | 4156 | UTR 3 | XP_011543336.1 | |||
XM_011545035.2 | 4156 | UTR 3 | XP_011543337.1 | |||
XM_017017750.1 | 4156 | UTR 3 | XP_016873239.1 | |||
XM_017017751.1 | 4156 | UTR 3 | XP_016873240.1 | |||
XM_017017752.1 | 4156 | UTR 3 | XP_016873241.1 | |||
XM_017017753.1 | 4156 | UTR 3 | XP_016873242.1 | |||
XM_017017754.1 | 4156 | UTR 3 | XP_016873243.1 |