Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HESX1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HESX1 - HESX homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003865.2 | 59 | UTR 5 | NP_003856.1 | |||
XM_005265526.4 | 59 | Intron | XP_005265583.1 | |||
XM_006713379.3 | 59 | Intron | XP_006713442.1 | |||
XM_011534204.2 | 59 | Intron | XP_011532506.1 | |||
XM_011534205.2 | 59 | Intron | XP_011532507.1 | |||
XM_017007421.1 | 59 | Intron | XP_016862910.1 |