Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMGCLL1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
HMGCLL1 - 3-hydroxymethyl-3-methylglutaryl-CoA lyase like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042406.1 | 2326 | UTR 3 | NP_001035865.1 | |||
NM_001287741.1 | 2326 | UTR 3 | NP_001274670.1 | |||
NM_001287746.1 | 2326 | UTR 3 | NP_001274675.1 | |||
NM_001287753.1 | 2326 | UTR 3 | NP_001274682.1 | |||
NM_019036.2 | 2326 | UTR 3 | NP_061909.2 | |||
XM_011514700.2 | 2326 | Intron | XP_011513002.1 |
Set Membership: |
HapMap |