Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600777 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TTF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TTF1 - transcription termination factor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001205296.1 | 3230 | Missense Mutation | GCG,GTG | A,V 370 | NP_001192225.1 | |
NM_007344.3 | 3230 | Missense Mutation | GCG,GTG | A,V 885 | NP_031370.2 | |
XM_006717273.3 | 3230 | Missense Mutation | GCG,GTG | A,V 885 | XP_006717336.2 | |
XM_017015105.1 | 3230 | Missense Mutation | GCG,GTG | A,V 918 | XP_016870594.1 |