Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCACCTGATTTTCAGAGGCCAGTT[C/G]CAGTGTTAGTATATGTAGTGGGAAA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603421 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MSANTD3-TMEFF1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MSANTD3-TMEFF1 - MSANTD3-TMEFF1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MURC - muscle related coiled-coil protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001018116.2 | Intron | NP_001018126.1 |
TMEFF1 - transmembrane protein with EGF like and two follistatin like domains 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |