Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604134 MIM: 613104 MIM: 606813 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAMTS13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
|||||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
ADAMTS13 - ADAM metallopeptidase with thrombospondin type 1 motif 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CACFD1 - calcium channel flower domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001135775.2 | 687 | Intron | NP_001129247.1 | |||
NM_001242369.1 | 687 | Intron | NP_001229298.1 | |||
NM_001242370.1 | 687 | Intron | NP_001229299.1 | |||
NM_017586.3 | 687 | Intron | NP_060056.1 | |||
XM_011518181.2 | 687 | UTR 5 | XP_011516483.1 | |||
XM_011518182.1 | 687 | Intron | XP_011516484.1 | |||
XM_011518184.1 | 687 | Intron | XP_011516486.1 | |||
XM_011518185.2 | 687 | UTR 5 | XP_011516487.1 | |||
XM_011518186.1 | 687 | Intron | XP_011516488.1 |
SLC2A6 - solute carrier family 2 member 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |