Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600709 MIM: 611534 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IARS PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IARS - isoleucyl-tRNA synthetase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3651 - microRNA 3651 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NOL8 - nucleolar protein 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256394.1 | 3783 | UTR 3 | NP_001243323.1 | |||
NM_017948.5 | 3783 | UTR 3 | NP_060418.4 | |||
XM_006717166.3 | 3783 | UTR 3 | XP_006717229.1 | |||
XM_006717167.3 | 3783 | UTR 3 | XP_006717230.1 | |||
XM_006717168.3 | 3783 | UTR 3 | XP_006717231.1 | |||
XM_006717169.3 | 3783 | UTR 3 | XP_006717232.1 | |||
XM_006717170.3 | 3783 | UTR 3 | XP_006717233.1 | |||
XM_006717172.3 | 3783 | UTR 3 | XP_006717235.1 | |||
XM_006717173.3 | 3783 | UTR 3 | XP_006717236.1 | |||
XM_011518824.2 | 3783 | UTR 3 | XP_011517126.1 | |||
XM_011518827.2 | 3783 | UTR 3 | XP_011517129.1 | |||
XM_017014876.1 | 3783 | UTR 3 | XP_016870365.1 | |||
XM_017014877.1 | 3783 | UTR 3 | XP_016870366.1 | |||
XM_017014878.1 | 3783 | UTR 3 | XP_016870367.1 | |||
XM_017014879.1 | 3783 | UTR 3 | XP_016870368.1 | |||
XM_017014880.1 | 3783 | UTR 3 | XP_016870369.1 |
SNORA84 - small nucleolar RNA, H/ACA box 84 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |