Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGCCAAAGAATTATATTGTTATTA[G/C]TAGCTAGAACCAAGATGTTGAAGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612246 MIM: 613334 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD302 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
|||||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
CD302 - CD302 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198763.1 | 8331 | UTR 3 | NP_001185692.1 | |||
NM_001198764.1 | 8331 | UTR 3 | NP_001185693.1 | |||
NM_014880.4 | 8331 | UTR 3 | NP_055695.2 |
LY75-CD302 - LY75-CD302 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198759.1 | 8331 | UTR 3 | NP_001185688.1 | |||
NM_001198760.1 | 8331 | UTR 3 | NP_001185689.1 |
MARCH7 - membrane associated ring-CH-type finger 7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282805.1 | 8331 | Intron | NP_001269734.1 | |||
NM_001282806.1 | 8331 | Intron | NP_001269735.1 | |||
NM_001282807.1 | 8331 | Intron | NP_001269736.1 | |||
NM_022826.3 | 8331 | Intron | NP_073737.1 | |||
XM_005246773.2 | 8331 | Intron | XP_005246830.1 | |||
XM_005246774.1 | 8331 | Intron | XP_005246831.1 | |||
XM_017004718.1 | 8331 | Intron | XP_016860207.1 | |||
XM_017004719.1 | 8331 | Intron | XP_016860208.1 | |||
XM_017004720.1 | 8331 | Intron | XP_016860209.1 | |||
XM_017004721.1 | 8331 | Intron | XP_016860210.1 |
Set Membership: |
HapMap Validated |