Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604047 MIM: 604048 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SLC22A13 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SLC22A13 - solute carrier family 22 member 13 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC22A14 - solute carrier family 22 member 14 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320033.1 | 796 | Intron | NP_001306962.1 | |||
NM_004803.4 | 796 | Intron | NP_004794.2 | |||
XM_005265585.4 | 796 | UTR 5 | XP_005265642.1 | |||
XM_006713416.3 | 796 | Intron | XP_006713479.2 | |||
XM_006713417.3 | 796 | Intron | XP_006713480.2 | |||
XM_006713418.3 | 796 | Intron | XP_006713481.2 | |||
XM_011534245.2 | 796 | UTR 5 | XP_011532547.1 | |||
XM_011534247.2 | 796 | Intron | XP_011532549.1 |