Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
48 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614698 MIM: 602869 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX20 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||||||||
EUR
|
|||||||||||
AMR
|
COX20 - COX20, cytochrome c oxidase assembly factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
HNRNPU - heterogeneous nuclear ribonucleoprotein U | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004501.3 | 6836 | UTR 3 | NP_004492.2 | |||
NM_031844.2 | 6836 | UTR 3 | NP_114032.2 | |||
XM_017001115.1 | 6836 | UTR 3 | XP_016856604.1 | |||
XM_017001116.1 | 6836 | UTR 3 | XP_016856605.1 | |||
XM_017001117.1 | 6836 | UTR 3 | XP_016856606.1 |
SNORA100 - small nucleolar RNA, H/ACA box 100 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap Validated |