Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
16 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 120831 MIM: 171835 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C4BPB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C4BPB - complement component 4 binding protein beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000716.3 | 67 | UTR 5 | NP_000707.1 | |||
NM_001017364.1 | 67 | UTR 5 | NP_001017364.1 | |||
NM_001017365.2 | 67 | Intron | NP_001017365.1 | |||
NM_001017366.2 | 67 | Intron | NP_001017366.1 | |||
NM_001017367.1 | 67 | UTR 5 | NP_001017367.1 | |||
XM_005273254.4 | 67 | UTR 5 | XP_005273311.1 | |||
XM_005273255.2 | 67 | Intron | XP_005273312.1 |
PFKFB2 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |