Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
57 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 103020 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AK2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
AK2 - adenylate kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199199.1 | 1660 | UTR 3 | NP_001186128.1 | |||
NM_001319139.1 | 1660 | UTR 3 | NP_001306068.1 | |||
NM_001319140.1 | 1660 | Intron | NP_001306069.1 | |||
NM_001319141.1 | 1660 | UTR 3 | NP_001306070.1 | |||
NM_001319142.1 | 1660 | UTR 3 | NP_001306071.1 | |||
NM_001319143.1 | 1660 | Intron | NP_001306072.1 | |||
NM_001625.3 | 1660 | Intron | NP_001616.1 | |||
NM_013411.4 | 1660 | UTR 3 | NP_037543.1 |
Set Membership: |
HapMap JSNP Validated |