Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
61 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606063 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
EXO1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
EXO1 - exonuclease 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319224.1 | 138 | Intron | NP_001306153.1 | |||
NM_003686.4 | 138 | Intron | NP_003677.4 | |||
NM_006027.4 | 138 | Intron | NP_006018.4 | |||
NM_130398.3 | 138 | Intron | NP_569082.2 | |||
XM_006711840.2 | 138 | Intron | XP_006711903.1 | |||
XM_011544321.1 | 138 | Intron | XP_011542623.1 | |||
XM_011544322.1 | 138 | UTR 5 | XP_011542624.1 | |||
XM_011544323.2 | 138 | Intron | XP_011542625.1 | |||
XM_011544324.2 | 138 | Intron | XP_011542626.1 | |||
XM_011544325.2 | 138 | Intron | XP_011542627.1 | |||
XM_017002793.1 | 138 | Intron | XP_016858282.1 |
Set Membership: |
HapMap |