Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 126090 MIM: 603729 | ||||||||||||||||||||
Literature Links: |
PCBD1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PCBD1 - pterin-4 alpha-carbinolamine dehydratase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000281.3 | Intron | NP_000272.1 | ||||
NM_001289797.1 | Intron | NP_001276726.1 | ||||
NM_001323004.1 | Intron | NP_001309933.1 |
SGPL1 - sphingosine-1-phosphate lyase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |