Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600483 MIM: 604039 MIM: 606456 | ||||||||||||||||||||
Literature Links: |
FGF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGF8 - fibroblast growth factor 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MGEA5 - meningioma expressed antigen 5 (hyaluronidase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142434.1 | 4120 | UTR 3 | NP_001135906.1 | |||
NM_012215.3 | 4120 | UTR 3 | NP_036347.1 | |||
XM_017015585.1 | 4120 | UTR 3 | XP_016871074.1 | |||
XM_017015586.1 | 4120 | UTR 3 | XP_016871075.1 |
NPM3 - nucleophosmin/nucleoplasmin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |