Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 136470 | ||||||||||||||||||||
Literature Links: |
FST PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FST - follistatin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006350.3 | 876 | Silent Mutation | GAA,GAG | E,E 236 | NP_006341.1 | |
NM_013409.2 | 876 | Silent Mutation | GAA,GAG | E,E 236 | NP_037541.1 | |
XM_005248400.4 | 876 | Silent Mutation | GAA,GAG | E,E 236 | XP_005248457.1 | |
XM_005248401.4 | 876 | Silent Mutation | GAA,GAG | E,E 236 | XP_005248458.1 | |
XM_005248402.4 | 876 | Silent Mutation | GAA,GAG | E,E 236 | XP_005248459.1 | |
XM_005248403.4 | 876 | Silent Mutation | GAA,GAG | E,E 236 | XP_005248460.1 | |
XM_011543099.2 | 876 | Silent Mutation | GAA,GAG | E,E 231 | XP_011541401.1 | |
XM_017008954.1 | 876 | Silent Mutation | GAA,GAG | E,E 236 | XP_016864443.1 | |
XM_017008955.1 | 876 | Silent Mutation | GAA,GAG | E,E 158 | XP_016864444.1 |