Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602338 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FAM217A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FAM217A - family with sequence similarity 217 member A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_173563.2 | 1916 | Missense Mutation | ATC,GTC | I,V 431 | NP_775834.2 | |
XM_005248951.3 | 1916 | Missense Mutation | ATC,GTC | I,V 422 | XP_005249008.1 | |
XM_006715025.2 | 1916 | Missense Mutation | ATC,GTC | I,V 368 | XP_006715088.1 | |
XM_006715026.3 | 1916 | Missense Mutation | ATC,GTC | I,V 368 | XP_006715089.1 | |
XM_011514413.2 | 1916 | Missense Mutation | ATC,GTC | I,V 368 | XP_011512715.1 | |
XM_011514414.2 | 1916 | Missense Mutation | ATC,GTC | I,V 368 | XP_011512716.1 | |
XM_011514415.1 | 1916 | Missense Mutation | ATC,GTC | I,V 421 | XP_011512717.1 | |
XM_011514416.2 | 1916 | Missense Mutation | ATC,GTC | I,V 418 | XP_011512718.1 | |
XM_011514417.1 | 1916 | Missense Mutation | ATC,GTC | I,V 383 | XP_011512719.1 | |
XM_017010480.1 | 1916 | Missense Mutation | ATC,GTC | I,V 451 | XP_016865969.1 | |
XM_017010481.1 | 1916 | Missense Mutation | ATC,GTC | I,V 422 | XP_016865970.1 | |
XM_017010482.1 | 1916 | Missense Mutation | ATC,GTC | I,V 368 | XP_016865971.1 |
PRPF4B - pre-mRNA processing factor 4B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |