Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTATTACAATTTCCTGACCCAGCT[A/G]TCTGGCCCTGGGCCAGCGTCTTAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RNF144A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RNF144A - ring finger protein 144A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014746.4 | Intron | NP_055561.2 | ||||
XM_005246200.3 | Intron | XP_005246257.1 | ||||
XM_005246202.4 | Intron | XP_005246259.1 | ||||
XM_017005396.1 | Intron | XP_016860885.1 | ||||
XM_017005397.1 | Intron | XP_016860886.1 | ||||
XM_017005398.1 | Intron | XP_016860887.1 | ||||
XM_017005399.1 | Intron | XP_016860888.1 | ||||
XM_017005400.1 | Intron | XP_016860889.1 | ||||
XM_017005401.1 | Intron | XP_016860890.1 | ||||
XM_017005402.1 | Intron | XP_016860891.1 | ||||
XM_017005403.1 | Intron | XP_016860892.1 | ||||
XM_017005404.1 | Intron | XP_016860893.1 |
RNF144A-AS1 - RNF144A antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |