Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613922 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LIPK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LIPK - lipase family member K | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080518.1 | Intron | NP_001073987.1 | ||||
XM_011540073.2 | Intron | XP_011538375.1 | ||||
XM_011540075.1 | Intron | XP_011538377.1 | ||||
XM_011540077.1 | Intron | XP_011538379.1 | ||||
XM_011540082.1 | Intron | XP_011538384.1 | ||||
XM_017016545.1 | Intron | XP_016872034.1 | ||||
XM_017016546.1 | Intron | XP_016872035.1 | ||||
XM_017016547.1 | Intron | XP_016872036.1 | ||||
XM_017016548.1 | Intron | XP_016872037.1 |