Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607559 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANKS3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ANKS3 - ankyrin repeat and sterile alpha motif domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242929.1 | 2014 | UTR 3 | NP_001229858.1 | |||
NM_001308089.1 | 2014 | UTR 3 | NP_001295018.1 | |||
NM_001324129.1 | 2014 | UTR 3 | NP_001311058.1 | |||
NM_001324130.1 | 2014 | UTR 3 | NP_001311059.1 | |||
NM_133450.3 | 2014 | UTR 3 | NP_597707.1 | |||
XM_005255119.3 | 2014 | UTR 3 | XP_005255176.1 | |||
XM_005255121.3 | 2014 | UTR 3 | XP_005255178.1 | |||
XM_011522372.1 | 2014 | UTR 3 | XP_011520674.1 | |||
XM_011522373.1 | 2014 | UTR 3 | XP_011520675.1 | |||
XM_011522374.1 | 2014 | UTR 3 | XP_011520676.1 | |||
XM_011522378.1 | 2014 | UTR 3 | XP_011520680.1 | |||
XM_017022935.1 | 2014 | UTR 3 | XP_016878424.1 | |||
XM_017022936.1 | 2014 | UTR 3 | XP_016878425.1 |
MGRN1 - mahogunin ring finger 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NUDT16L1 - nudix hydrolase 16 like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193452.1 | 2014 | Intron | NP_001180381.1 | |||
NM_032349.3 | 2014 | Intron | NP_115725.1 | |||
XM_005255633.4 | 2014 | Intron | XP_005255690.1 |