Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606280 | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DCAF16 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Global
|
Caucasian
|
CEPH (CEU)
|
|||||||||
EAS
|
African American
|
YRI (Yoruba)
|
|||||||||
SAS
|
Japanese
|
JPT (Japanese)
|
|||||||||
AFR
|
Chinese
|
CHB (Han Chinese)
|
|||||||||
EUR
|
|||||||||||
AMR
|
DCAF16 - DDB1 and CUL4 associated factor 16 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NCAPG - non-SMC condensin I complex subunit G | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_022346.4 | Intron | NP_071741.2 | ||||
XM_011513876.2 | Intron | XP_011512178.1 | ||||
XM_017008543.1 | Intron | XP_016864032.1 | ||||
XM_017008544.1 | Intron | XP_016864033.1 |