Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 604261 | ||||||||||||||||||||
Literature Links: |
ATG5 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
ATG5 - autophagy related 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286106.1 | 2572 | UTR 3 | NP_001273035.1 | |||
NM_001286107.1 | 2572 | UTR 3 | NP_001273036.1 | |||
NM_001286108.1 | 2572 | UTR 3 | NP_001273037.1 | |||
NM_001286111.1 | 2572 | UTR 3 | NP_001273040.1 | |||
NM_004849.3 | 2572 | UTR 3 | NP_004840.1 |