Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGTCCCCCATGCAGACGCGGTTC[C/T]ACTCGCAGCCACAGTTCCAGCACCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602544 | ||||||||||||||||||||
Literature Links: |
PARK2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PARK2 - parkin RBR E3 ubiquitin protein ligase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004562.2 | 1489 | Nonsense Mutation | TAG,TGG | *,W 453 | NP_004553.2 | |
NM_013987.2 | 1489 | Nonsense Mutation | TAG,TGG | *,W 425 | NP_054642.2 | |
NM_013988.2 | 1489 | Nonsense Mutation | TAG,TGG | *,W 304 | NP_054643.2 | |
XM_011535863.1 | 1489 | Nonsense Mutation | TAG,TGG | *,W 452 | XP_011534165.1 | |
XM_017010908.1 | 1489 | Nonsense Mutation | TAG,TGG | *,W 491 | XP_016866397.1 | |
XM_017010909.1 | 1489 | Nonsense Mutation | TAG,TGG | *,W 373 | XP_016866398.1 |