Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607922 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
A4GALT PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
A4GALT - alpha 1,4-galactosyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318038.1 | 2185 | UTR 3 | NP_001304967.1 | |||
NM_017436.5 | 2185 | UTR 3 | NP_059132.1 | |||
XM_005261644.2 | 2185 | UTR 3 | XP_005261701.1 | |||
XM_005261646.4 | 2185 | UTR 3 | XP_005261703.1 | |||
XM_005261647.2 | 2185 | UTR 3 | XP_005261704.1 | |||
XM_005261648.3 | 2185 | UTR 3 | XP_005261705.1 | |||
XM_006724265.3 | 2185 | UTR 3 | XP_006724328.1 | |||
XM_006724266.3 | 2185 | UTR 3 | XP_006724329.1 | |||
XM_011530233.2 | 2185 | UTR 3 | XP_011528535.1 | |||
XM_017028831.1 | 2185 | UTR 3 | XP_016884320.1 |
Set Membership: |
HapMap |