Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCGTGTTTTCCCACAGTCTGTCTA[C/T]GGTCCATTCACCATCTGTGAGTGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BANF2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BANF2 - barrier to autointegration factor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001014977.3 | Intron | NP_001014977.2 | ||||
NM_001159495.1 | Intron | NP_001152967.1 | ||||
NM_178477.4 | Intron | NP_848572.3 | ||||
XM_005260668.3 | Intron | XP_005260725.1 | ||||
XM_011529170.1 | Intron | XP_011527472.1 | ||||
XM_011529171.2 | Intron | XP_011527473.1 |
LOC105372547 - uncharacterized LOC105372547 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |